Learn More
Meso Scale Discovery U-PLEX Obesity Combo 2 ms
SDP
Catalog No. 5000082246
Encompass
The Meso Scale Discovery U-PLEX Obesity Combo 2 Mouse Assay is a multiplex immunoassay panel that measures biomarkers related to obesity and metabolic disorders in mouse samples The assay simultaneously measures leptin insulin resistin and PAI-1 in 50 L of sample The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay requires the Meso Scale Discovery SECTOR Imager and provides components for one 96-well plate It is intended for research use only
Catalog No. 50-000-82246 Supplier Meso Scale Discovery Supplier No. K15301K1
The Meso Scale Discovery U-PLEX Obesity Combo 2 Mouse Assay is a multiplex immunoassay panel that measures biomarkers related to obesity and metabolic disorders in mouse samples The assay simultaneously measures leptin insulin resistin and PAI-1 in 50 L of sample The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This assay requires the Meso Scale Discovery SECTOR Imager and provides components for one 96-well plate It is intended for research use only