Learn More
Meso Scale Discovery U-PLEX Obesity Combo 2 ms
SDP
Catalog No. 5000082247
Encompass
The U-PLEX Obesity Combo 2 Mouse is a multiplex assay kit from Meso Scale Diagnostics for measuring 10 obesity-related biomarkers in mouse samples It uses electrochemiluminescent detection and tests 75 samples in triplicate The panel includes leptin insulin PAI-1 resistin TNF-alpha MCP-1 IL-6 adiponectin amylin and glucagon This kit is ideal for profiling biomarkers linked to obesity inflammation insulin resistance and diabetes in mouse models offering high sensitivity broad dynamic range and low sample volume requirements for research
Catalog No. 50-000-82247 Supplier Meso Scale Discovery Supplier No. K15301K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Obesity Combo 2 Mouse is a multiplex assay kit from Meso Scale Diagnostics for measuring 10 obesity-related biomarkers in mouse samples It uses electrochemiluminescent detection and tests 75 samples in triplicate The panel includes leptin insulin PAI-1 resistin TNF-alpha MCP-1 IL-6 adiponectin amylin and glucagon This kit is ideal for profiling biomarkers linked to obesity inflammation insulin resistance and diabetes in mouse models offering high sensitivity broad dynamic range and low sample volume requirements for research