Learn More
Meso Scale Discovery U-PLEX Obesity Combo 2 rt
SDP
Catalog No. 5000082389
Encompass
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassay panels for detecting biomarkers cytokines chemokines growth factors and immunoglobulins from a single sample Using electrochemiluminescence technology they target various disease states and biomarker research U-PLEX panels offer high sensitivity broad dynamic range and low sample volume requirements Available in a 96-well plate format they work with serum plasma and cell culture supernatants The assays require no wash steps and deliver results in 3 5 hours
Catalog No. 50-000-82389 Supplier Meso Scale Discovery Supplier No. K15311K1
The Meso Scale Discovery U-PLEX Combinations are multiplex immunoassay panels for detecting biomarkers cytokines chemokines growth factors and immunoglobulins from a single sample Using electrochemiluminescence technology they target various disease states and biomarker research U-PLEX panels offer high sensitivity broad dynamic range and low sample volume requirements Available in a 96-well plate format they work with serum plasma and cell culture supernatants The assays require no wash steps and deliver results in 3 5 hours