Learn More
Meso Scale Discovery U-PLEX Proinflam Combo 2 hu
SDP
Catalog No. 5000080576
Encompass
The Meso Scale Discovery U-PLEX ProInflam Combo 2 Human Kit is a multiplex immunoassay measuring human IFN-gamma IL-10 IL-12p70 IL-13 IL-1beta IL-2 IL-4 IL-6 IL-8 and TNF-alpha in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 7-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies buffers diluent calibrators and controls enabling efficient profiling of multiple inflammatory biomarkers from a small sample
Catalog No. 50-000-80576 Supplier Meso Scale Discovery Supplier No. K15066K1
The Meso Scale Discovery U-PLEX ProInflam Combo 2 Human Kit is a multiplex immunoassay measuring human IFN-gamma IL-10 IL-12p70 IL-13 IL-1beta IL-2 IL-4 IL-6 IL-8 and TNF-alpha in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 7-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies buffers diluent calibrators and controls enabling efficient profiling of multiple inflammatory biomarkers from a small sample