Learn More
Meso Scale Discovery U-PLEX Proinflam Combo 2 hu
SDP
Catalog No. 5000081697
Encompass
The U-PLEX ProInflam Combo 2 Human is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of 10 human inflammatory biomarkers (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- ) in serum plasma and culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes all necessary reagents and plates for running the assay
Catalog No. 50-000-81697 Supplier Meso Scale Discovery Supplier No. K15066K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX ProInflam Combo 2 Human is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of 10 human inflammatory biomarkers (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- ) in serum plasma and culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes all necessary reagents and plates for running the assay