Learn More
Meso Scale Discovery U-PLEX Proinflam Combo 3 hu
SDP
Catalog No. 5000080520
Encompass
The Meso Scale Discovery U-PLEX ProInflam Combo 3 Human Kit is a multiplex immunoassay that simultaneously measures IL-6 IL-8 and TNF-alpha in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It can quantitatively determine cytokine levels with accuracy and reproducibility This assay kit enables efficient profiling of multiple inflammatory biomarkers from a single small-volume sample
Catalog No. 50-000-80520 Supplier Meso Scale Discovery Supplier No. K15052K1
The Meso Scale Discovery U-PLEX ProInflam Combo 3 Human Kit is a multiplex immunoassay that simultaneously measures IL-6 IL-8 and TNF-alpha in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer It can quantitatively determine cytokine levels with accuracy and reproducibility This assay kit enables efficient profiling of multiple inflammatory biomarkers from a single small-volume sample