Learn More
Meso Scale Discovery U-PLEX Proinflam Combo 4 (human), multiplex assay kit, 5 plates, detects IL-1β, IL-6, IL-8, TNF-α, 96-well plate format
SDP
Catalog No. NC3444076
Encompass
U-PLEX Proinflam Combo 4 (human), multiplex assay kit, 5 plates, detects IL-1β, IL-6, IL-8, TNF-α, 96-well plate format
Catalog No. NC3444076 Supplier Meso Scale Discovery Supplier No. K15053K2
U-PLEX Proinflam Combo 4 (human), multiplex assay kit, 5 plates, detects IL-1β, IL-6, IL-8, TNF-α, 96-well plate format