Learn More
Meso Scale Discovery U-PLEX Proinflam Combo 4 hu
SDP
Catalog No. 5000078771
Encompass
The Meso Scale Discovery U-PLEX ProInflam Combo 4 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 4 human proinflammatory biomarkers IFN- IL-1 IL-6 and TNF- in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s patented MULTI-ARRAY technology and is run on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay allows high-throughput sensitive and reproducible quantification of multiple inflammatory biomarkers from a small sample volume
Catalog No. 50-000-78771 Supplier Meso Scale Discovery Supplier No. K15053K1
The Meso Scale Discovery U-PLEX ProInflam Combo 4 Human Kit is a multiplex immunoassay that simultaneously measures the levels of 4 human proinflammatory biomarkers IFN- IL-1 IL-6 and TNF- in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s patented MULTI-ARRAY technology and is run on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay allows high-throughput sensitive and reproducible quantification of multiple inflammatory biomarkers from a small sample volume