Learn More
Meso Scale Discovery U-PLEX Rat C-Peptide ASY
SDP
Catalog No. 5000081362
Encompass
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for quantifying C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It offers high specificity for rat C-peptide with minimal cross-reactivity The standard curve range is 50-50 000 pg/mL with a minimum detectable dose of 17 pg/mL
Catalog No. 50-000-81362 Supplier Meso Scale Discovery Supplier No. K1536JK2
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for quantifying C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It offers high specificity for rat C-peptide with minimal cross-reactivity The standard curve range is 50-50 000 pg/mL with a minimum detectable dose of 17 pg/mL