Learn More
Meso Scale Discovery U-PLEX Rat C-Peptide ASY
SDP
Catalog No. 5000081914
Encompass
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay that quantitatively measures C-peptide in rat serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay C-peptide levels can be quantified in less than 3 5 hours This product allows for the accurate measurement of C-peptide which can aid in studies of insulin secretion and diabetes
Catalog No. 50-000-81914 Supplier Meso Scale Discovery Supplier No. K1536JK1
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay that quantitatively measures C-peptide in rat serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay C-peptide levels can be quantified in less than 3 5 hours This product allows for the accurate measurement of C-peptide which can aid in studies of insulin secretion and diabetes