Learn More
Meso Scale Discovery U-PLEX Rat C-Peptide ASYQP
SDP
Catalog No. 5000080345
Encompass
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for measuring C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and diluent Sensitivity is 1 3 pg/mL with a quantification limit of 10 000 pg/mL Results are available in 3 5 hours using an MSD instrument like the MESO QuickPlex SQ 120
Catalog No. 50-000-80345 Supplier Meso Scale Discovery Supplier No. K1536JK21
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for measuring C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and diluent Sensitivity is 1 3 pg/mL with a quantification limit of 10 000 pg/mL Results are available in 3 5 hours using an MSD instrument like the MESO QuickPlex SQ 120