Learn More
Meso Scale Discovery U-PLEX Rat C-Peptide ASYQP
SDP
Catalog No. 5000081363
Encompass
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for measuring C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and an MSD Read Buffer Results are ready in 3 5 hours using an MSD instrument like MESO QuickPlex SQ 120 Detection limits range from 1 5 pg/mL to 10 000 pg/mL
Catalog No. 50-000-81363 Supplier Meso Scale Discovery Supplier No. K1536JK22
The Meso Scale Discovery U-PLEX Rat C-Peptide Assay Kit is a multiplex immunoassay for measuring C-peptide in rat serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity and a wide dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators and an MSD Read Buffer Results are ready in 3 5 hours using an MSD instrument like MESO QuickPlex SQ 120 Detection limits range from 1 5 pg/mL to 10 000 pg/mL