Learn More
Meso Scale Discovery U-PLEX Rat FGF-21 ASY 25 PL
SDP
Catalog No. 5000081940
Encompass
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is a multiplex immunoassay that measures FGF-21 levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrator and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It can be used to measure FGF-21 levels in studies related to metabolism diabetes obesity and mitochondrial disorders
Catalog No. 50-000-81940 Supplier Meso Scale Discovery Supplier No. K1535WK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is a multiplex immunoassay that measures FGF-21 levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrator and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It can be used to measure FGF-21 levels in studies related to metabolism diabetes obesity and mitochondrial disorders