Learn More
Meso Scale Discovery U-PLEX Rat FGF-21 ASY
SDP
Catalog No. 5000078268
Encompass
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is a multiplex immunoassay that measures FGF-21 levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It allows for the quantitative determination of FGF-21 in less than 3 5 hours
Catalog No. 50-000-78268 Supplier Meso Scale Discovery Supplier No. K1535WK1
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is a multiplex immunoassay that measures FGF-21 levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It allows for the quantitative determination of FGF-21 in less than 3 5 hours