Learn More
Meso Scale Discovery U-PLEX Rat FGF-21 ASY
SDP
Supplier:  Meso Scale Discovery K1535WK2
Encompass
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of FGF-21 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology coated with capture antibodies and SULFO-TAG labeled detection antibodies to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate detection antibody calibrator and assay diluent valid for 50 tests The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81889