Learn More
Meso Scale Discovery U-PLEX Rat FGF-21 ASY384-well
SDP
Catalog No. 5000081220
Encompass
The Meso Scale Discovery U-PLEX Rat FGF-21 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of FGF-21 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD Streptavidin Multi-Array plates and a proprietary ruthenium labeled detection antibody to provide high sensitivity and a wide dynamic range The kit includes a calibrator to generate a standard curve and reagents for up to 384 tests in a 384-well plate format The streamlined no-wash protocol provides results in under 3 hours The assay has high sensitivity with an LLOQ of 1 15 pg/mL and requires as little as 25 L of sample
Catalog No. 50-000-81220 Supplier Meso Scale Discovery Supplier No. K2535WK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat FGF-21 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of FGF-21 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD Streptavidin Multi-Array plates and a proprietary ruthenium labeled detection antibody to provide high sensitivity and a wide dynamic range The kit includes a calibrator to generate a standard curve and reagents for up to 384 tests in a 384-well plate format The streamlined no-wash protocol provides results in under 3 hours The assay has high sensitivity with an LLOQ of 1 15 pg/mL and requires as little as 25 L of sample