Learn More
Meso Scale Discovery U-PLEX Rat FGF-21 ASYQP
SDP
Catalog No. 5000079395
Encompass
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is an electrochemiluminescent immunoassay for quantifying FGF-21 in rat serum plasma and tissue culture media It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and buffers Sensitivity is 1 3 pg/mL with an upper limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours Ideal for research in biomarker discovery and disease
Catalog No. 50-000-79395 Supplier Meso Scale Discovery Supplier No. K1535WK22
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is an electrochemiluminescent immunoassay for quantifying FGF-21 in rat serum plasma and tissue culture media It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and buffers Sensitivity is 1 3 pg/mL with an upper limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours Ideal for research in biomarker discovery and disease