Learn More
Meso Scale Discovery U-PLEX Rat FGF-21 ASYQP
SDP
Catalog No. 5000079848
Encompass
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is a multiplex immunoassay for measuring FGF-21 in rat serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It s suitable for studies on metabolism diabetes obesity and mitochondrial disorders offering high performance and reliability
Catalog No. 50-000-79848 Supplier Meso Scale Discovery Supplier No. K1535WK21
The Meso Scale Discovery U-PLEX Rat FGF-21 Assay Kit is a multiplex immunoassay for measuring FGF-21 in rat serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL It s suitable for studies on metabolism diabetes obesity and mitochondrial disorders offering high performance and reliability