Learn More
Meso Scale Discovery U-PLEX Rat Ghrelin total ASY
SDP
Catalog No. 5000079374
Encompass
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay for measuring total ghrelin in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for simultaneous detection in a single well The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer It has a sensitivity of 1 3 pg/mL and an upper limit of 10 000 pg/mL offering high sensitivity low sample volume and a 2 5-hour protocol
Catalog No. 50-000-79374 Supplier Meso Scale Discovery Supplier No. K1535XK2
The Meso Scale Discovery U-PLEX Rat Ghrelin Total Assay Kit is a multiplex immunoassay for measuring total ghrelin in rat serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for simultaneous detection in a single well The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer It has a sensitivity of 1 3 pg/mL and an upper limit of 10 000 pg/mL offering high sensitivity low sample volume and a 2 5-hour protocol