Learn More
Meso Scale Discovery U-PLEX Rat GLP-1 active ASY
SDP
Catalog No. 5000081500
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit quantifies active GLP-1 in rat serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a dynamic range of up to 4 logs and low sample volume needs offering a robust tool for studying active GLP-1 in preclinical rat models
Catalog No. 50-000-81500 Supplier Meso Scale Discovery Supplier No. K1536LK2
The Meso Scale Discovery U-PLEX Rat GLP-1 (Active) Assay Kit quantifies active GLP-1 in rat serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive rapid detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a dynamic range of up to 4 logs and low sample volume needs offering a robust tool for studying active GLP-1 in preclinical rat models