Learn More
Meso Scale Discovery U-PLEX Rat GLP-1 total ASY
SDP
Catalog No. 5000079391
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 15 pg/mL and the upper limit of quantitation is 10 000 pg/mL The assay measures both intact GLP-1 and the metabolite GLP-1 9-36amide It is for research use only
Catalog No. 50-000-79391 Supplier Meso Scale Discovery Supplier No. K1535UK1
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 15 pg/mL and the upper limit of quantitation is 10 000 pg/mL The assay measures both intact GLP-1 and the metabolite GLP-1 9-36amide It is for research use only