Learn More
Meso Scale Discovery U-PLEX Rat GLP-1 total ASY
SDP
Catalog No. 5000081887
Encompass
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity wide dynamic range and low sample volume requirements This multiplex assay allows simultaneous measurement of GLP-1 and other biomarkers The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay
Catalog No. 50-000-81887 Supplier Meso Scale Discovery Supplier No. K1535UK2
The Meso Scale Discovery U-PLEX Rat GLP-1 Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total GLP-1 in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity wide dynamic range and low sample volume requirements This multiplex assay allows simultaneous measurement of GLP-1 and other biomarkers The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay