Learn More
Meso Scale Discovery U-PLEX Rat GLP-1 total ASYQP
SDP
Catalog No. 5000079392
Encompass
The Meso Scale Discovery GLP-1 assay kit is a sensitive electrochemiluminescent immunoassay for measuring active GLP-1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology to detect GLP-1 at sub-picogram per milliliter levels with high specificity The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents The assay takes 3 5 hours enabling sensitive reproducible quantification of active GLP-1 for research
Catalog No. 50-000-79392 Supplier Meso Scale Discovery Supplier No. K1535UK22
The Meso Scale Discovery GLP-1 assay kit is a sensitive electrochemiluminescent immunoassay for measuring active GLP-1 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology to detect GLP-1 at sub-picogram per milliliter levels with high specificity The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents The assay takes 3 5 hours enabling sensitive reproducible quantification of active GLP-1 for research