Learn More
Meso Scale Discovery U-PLEX Rat GLP-1 total ASYQP
SDP
Catalog No. 5000081888
Encompass
The Meso Scale Discovery GLP-1 assay kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in human serum plasma and other biological samples The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to enable rapid and accurate detection of GLP-1 levels down to the pg/mL range This assay kit provides a fast reliable method for quantifying active GLP-1 which can aid in the study of diabetes obesity and metabolic disorders The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and all necessary reagents for the immunoassay
Catalog No. 50-000-81888 Supplier Meso Scale Discovery Supplier No. K1535UK21
The Meso Scale Discovery GLP-1 assay kit is a highly sensitive electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in human serum plasma and other biological samples The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to enable rapid and accurate detection of GLP-1 levels down to the pg/mL range This assay kit provides a fast reliable method for quantifying active GLP-1 which can aid in the study of diabetes obesity and metabolic disorders The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and all necessary reagents for the immunoassay