Learn More
Meso Scale Discovery U-PLEX Rat Glucagon ASYQP
SDP
Catalog No. 5000081941
Encompass
The Meso Scale Discovery U-PLEX Rat Glucagon Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 15 pg/mL and the upper limit of quantitation is 10 000 pg/mL The assay takes approximately 3 5 hours to complete and includes a 96-well plate coated with capture antibody detection antibody and read buffer for electrochemiluminescent signal generation on the MSD instrument platform
Catalog No. 50-000-81941 Supplier Meso Scale Discovery Supplier No. K1535YK21
The Meso Scale Discovery U-PLEX Rat Glucagon Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in rat serum plasma and cell culture supernatants The assay uses MSD GOLD plate technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 25 L The lower limit of detection is 1 15 pg/mL and the upper limit of quantitation is 10 000 pg/mL The assay takes approximately 3 5 hours to complete and includes a 96-well plate coated with capture antibody detection antibody and read buffer for electrochemiluminescent signal generation on the MSD instrument platform