Learn More
Meso Scale Discovery U-PLEX Rat Insulin ASY384-well
SDP
Catalog No. 5000081158
Encompass
The Meso Scale Discovery U-PLEX Rat Insulin 384-Well Assay Kit is a multiplex immunoassay that measures insulin levels in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of samples The assay has a detection range of 50-50 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator containing recombinant rat insulin The assay takes approximately 3 hours to complete and 10 L of sample is required
Catalog No. 50-000-81158 Supplier Meso Scale Discovery Supplier No. K2536HK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Insulin 384-Well Assay Kit is a multiplex immunoassay that measures insulin levels in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput analysis of samples The assay has a detection range of 50-50 000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator containing recombinant rat insulin The assay takes approximately 3 hours to complete and 10 L of sample is required