Learn More
Meso Scale Discovery U-PLEX Rat Leptin ASY 25 PL
SDP
Catalog No. 5000082083
Encompass
The Meso Scale Discovery U-PLEX Rat Leptin Assay Kit is a multiplex immunoassay that measures leptin levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for leptin
Catalog No. 50-000-82083 Supplier Meso Scale Discovery Supplier No. K1535ZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Rat Leptin Assay Kit is a multiplex immunoassay that measures leptin levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for leptin