Learn More
Meso Scale Discovery U-PLEX Rat Leptin ASY
SDP
Catalog No. 5000081942
Encompass
The Meso Scale Discovery U-PLEX Rat Leptin Assay Kit is a multiplex immunoassay that measures leptin levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and controls The assay requires a small sample volume and provides results in as little as 3 5 hours With minimal hands-on time this kit allows for the rapid accurate quantification of rat leptin
Catalog No. 50-000-81942 Supplier Meso Scale Discovery Supplier No. K1535ZK1
The Meso Scale Discovery U-PLEX Rat Leptin Assay Kit is a multiplex immunoassay that measures leptin levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and controls The assay requires a small sample volume and provides results in as little as 3 5 hours With minimal hands-on time this kit allows for the rapid accurate quantification of rat leptin