Learn More
Meso Scale Discovery U-PLEX Rat Leptin ASYQP
SDP
Catalog No. 5000079797
Encompass
The Meso Scale Discovery U-PLEX Rat Leptin Assay Kit is a multiplex immunoassay that measures leptin levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 9 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79797 Supplier Meso Scale Discovery Supplier No. K1535ZK22
The Meso Scale Discovery U-PLEX Rat Leptin Assay Kit is a multiplex immunoassay that measures leptin levels in rat serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 9 pg/mL and an upper limit of quantification of 10 000 pg/mL