Learn More
Meso Scale Discovery U-PLEX Rat PYY total ASY
SDP
Catalog No. 5000078567
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay kit for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay allows for the simultaneous measurement of total PYY along with other biomarkers in the rat system The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The total assay time is 3 5 hours
Catalog No. 50-000-78567 Supplier Meso Scale Discovery Supplier No. K1536BK2
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay kit for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay allows for the simultaneous measurement of total PYY along with other biomarkers in the rat system The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The total assay time is 3 5 hours