Learn More
Meso Scale Discovery U-PLEX Rat PYY total ASY
SDP
Catalog No. 5000079760
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay that measures total PYY in rat samples The kit uses electrochemiluminescence detection technology and provides reagents for testing up to 1536 samples in a 96-well plate format The assay has a typical sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL The kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators controls assay and wash buffers and an MSD instrument for detection The assay requires a low sample volume of 50 L and has high specificity with minimal cross-reactivity
Catalog No. 50-000-79760 Supplier Meso Scale Discovery Supplier No. K1536BK1
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is a multiplex immunoassay that measures total PYY in rat samples The kit uses electrochemiluminescence detection technology and provides reagents for testing up to 1536 samples in a 96-well plate format The assay has a typical sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL The kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators controls assay and wash buffers and an MSD instrument for detection The assay requires a low sample volume of 50 L and has high specificity with minimal cross-reactivity