Learn More
Meso Scale Discovery U-PLEX Rat PYY total ASYQP
SDP
Catalog No. 5000079371
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates allowing detection of less than 1 pg/mL PYY with a 3 hour protocol This immunoassay provides a rapid ultra-sensitive method to measure total PYY levels for the study of appetite regulation and gastrointestinal function in rat models
Catalog No. 50-000-79371 Supplier Meso Scale Discovery Supplier No. K1536BK21
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates allowing detection of less than 1 pg/mL PYY with a 3 hour protocol This immunoassay provides a rapid ultra-sensitive method to measure total PYY levels for the study of appetite regulation and gastrointestinal function in rat models