Learn More
Meso Scale Discovery U-PLEX Rat PYY total ASYQP
SDP
Catalog No. 5000081938
Encompass
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide superior sensitivity and a broad dynamic range This kit includes a plate detection antibody calibrator and diluent valid for 50 tests when using recommended protocols The lower limit of detection is 1 5 pg/mL with an upper limit of 50 000 pg/mL
Catalog No. 50-000-81938 Supplier Meso Scale Discovery Supplier No. K1536BK22
The Meso Scale Discovery U-PLEX Rat PYY Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in rat serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide superior sensitivity and a broad dynamic range This kit includes a plate detection antibody calibrator and diluent valid for 50 tests when using recommended protocols The lower limit of detection is 1 5 pg/mL with an upper limit of 50 000 pg/mL