Learn More
Meso Scale Discovery U-PLEX T-Cell Combo ms
SDP
Catalog No. 5000078820
Encompass
The U-PLEX T Cell Combo Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous detection of mouse IFN-gamma IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IL-17A IL-17F IL-21 IL-22 IL-23 IL-27 IL-31 IL-33 TNF-alpha GM-CSF RANTES MIP-1alpha MIP-1beta MCP-1 IP-10 MIG Eotaxin TARC and MDC in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The electrochemiluminescent immunoassay allows for high sensitivity and a broad dynamic range
Catalog No. 50-000-78820 Supplier Meso Scale Discovery Supplier No. K15098K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX T Cell Combo Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for the simultaneous detection of mouse IFN-gamma IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IL-17A IL-17F IL-21 IL-22 IL-23 IL-27 IL-31 IL-33 TNF-alpha GM-CSF RANTES MIP-1alpha MIP-1beta MCP-1 IP-10 MIG Eotaxin TARC and MDC in cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The electrochemiluminescent immunoassay allows for high sensitivity and a broad dynamic range