Learn More
Meso Scale Discovery U-PLEX T-Cell Combo NHP
SDP
Catalog No. 5000081764
Encompass
The U-PLEX T Cell Combo NHP Kit from Meso Scale Diagnostics is a multiplex assay kit for measuring monkey interferon gamma interleukin 2 4 5 10 and 17A in cell culture supernatants and serum This electrochemiluminescence immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium buffers serum diluent and cytokine standards for calibration curves offering a streamlined workflow and rapid results for NHP samples
Catalog No. 50-000-81764 Supplier Meso Scale Discovery Supplier No. K15095K1
The U-PLEX T Cell Combo NHP Kit from Meso Scale Diagnostics is a multiplex assay kit for measuring monkey interferon gamma interleukin 2 4 5 10 and 17A in cell culture supernatants and serum This electrochemiluminescence immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium buffers serum diluent and cytokine standards for calibration curves offering a streamlined workflow and rapid results for NHP samples