Learn More
Meso Scale Discovery U-PLEX T-Cell Combo NHP
SDP
Catalog No. 5000081774
Encompass
The U-PLEX T Cell Combo NHP Kit from Meso Scale Diagnostics is a multiplex assay kit that allows for the simultaneous quantitative measurement of monkey interferon gamma interleukin 2 4 5 10 and 17A in serum plasma cell culture supernatants and other biological samples The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance The assay requires 50 L of sample and can be completed in under 5 hours
Catalog No. 50-000-81774 Supplier Meso Scale Discovery Supplier No. K15095K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX T Cell Combo NHP Kit from Meso Scale Diagnostics is a multiplex assay kit that allows for the simultaneous quantitative measurement of monkey interferon gamma interleukin 2 4 5 10 and 17A in serum plasma cell culture supernatants and other biological samples The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance The assay requires 50 L of sample and can be completed in under 5 hours