Learn More
Meso Scale Discovery U-PLEX TBI Combo 1 human
SDP
Catalog No. 5000079307
Encompass
The Meso Scale Discovery U-PLEX TBI Combo 1 Human Kit is a multiplex assay for measuring Tau GFAP and UCH-L1 biomarkers using electrochemiluminescence (ECL) technology It provides a sensitive and reproducible method for human serum plasma and CSF samples The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators controls diluent read buffer wash buffer and plate seals It has a range of 50-25 000 pg/mL offering high sensitivity and a broad dynamic range with low sample volume
Catalog No. 50-000-79307 Supplier Meso Scale Discovery Supplier No. K15685K1
The Meso Scale Discovery U-PLEX TBI Combo 1 Human Kit is a multiplex assay for measuring Tau GFAP and UCH-L1 biomarkers using electrochemiluminescence (ECL) technology It provides a sensitive and reproducible method for human serum plasma and CSF samples The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators controls diluent read buffer wash buffer and plate seals It has a range of 50-25 000 pg/mL offering high sensitivity and a broad dynamic range with low sample volume