Learn More
Meso Scale Discovery U-PLEX TBI Combo 1 human
SDP
Catalog No. 5000079308
Encompass
The Meso Scale Discovery U-PLEX TBI Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures biomarkers for traumatic brain injury (TBI) in human serum plasma and CSF samples The kit includes reagents to measure GFAP UCH-L1 S100B and Tau in a single well of a 96-well plate using electrochemiluminescence detection The assay has high sensitivity requires low sample volume and provides results in 3 5 hours This kit enables efficient TBI biomarker profiling to aid diagnosis and monitoring of mild to severe TBI
Catalog No. 50-000-79308 Supplier Meso Scale Discovery Supplier No. K15685K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX TBI Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures biomarkers for traumatic brain injury (TBI) in human serum plasma and CSF samples The kit includes reagents to measure GFAP UCH-L1 S100B and Tau in a single well of a 96-well plate using electrochemiluminescence detection The assay has high sensitivity requires low sample volume and provides results in 3 5 hours This kit enables efficient TBI biomarker profiling to aid diagnosis and monitoring of mild to severe TBI