Learn More
Meso Scale Discovery U-PLEX TGF-B Combo NHP
SDP
Catalog No. 5000082169
Encompass
The U-PLEX TGF-beta Combo NHP Assay Kit from Meso Scale Diagnostics measures active and total TGF-beta 1 2 and 3 in serum plasma and cell culture supernatants using MSD s electrochemiluminescent detection It offers high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture and detection antibodies calibrators and controls The assay runs in 3 5 hours and requires an MSD instrument like QuickPlex for readout enabling efficient quantification of TGF-beta isoforms in small sample volumes
Catalog No. 50-000-82169 Supplier Meso Scale Discovery Supplier No. K15243K1
The U-PLEX TGF-beta Combo NHP Assay Kit from Meso Scale Diagnostics measures active and total TGF-beta 1 2 and 3 in serum plasma and cell culture supernatants using MSD s electrochemiluminescent detection It offers high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture and detection antibodies calibrators and controls The assay runs in 3 5 hours and requires an MSD instrument like QuickPlex for readout enabling efficient quantification of TGF-beta isoforms in small sample volumes