Learn More
Meso Scale Discovery U-PLEX TGF-B Combo NHP
SDP
Catalog No. 5000082170
Encompass
The U-PLEX TGF-beta Combo NHP Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active and total TGF-beta 1 TGF-beta 2 and TGF-beta 3 in serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to run the assay Results are obtained in as little as 3 5 hours The assay is for research use only
Catalog No. 50-000-82170 Supplier Meso Scale Discovery Supplier No. K15243K2
The U-PLEX TGF-beta Combo NHP Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active and total TGF-beta 1 TGF-beta 2 and TGF-beta 3 in serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to run the assay Results are obtained in as little as 3 5 hours The assay is for research use only