Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery U-PLEX TH1/TH2 Combo Kit, 10 analytes, Human, Electrochemiluminescence (ECL), Sub- to low pg/mL range sensitivity, Cell culture supernatant/ serum/ plasma
SDP

Catalog No. NC3132558
Encompass

The MESO SCALE DISCOVERY U-PLEX TH1/TH2 Combo (human) is a pre-configured multiplex assay kit designed to measure human cytokines involved in inflammation and the Th1/Th2 pathways. This kit features advanced U-PLEX technology that enables the efficient and sensitive detection of multiple analytes in a single sample, making it an ideal choice for research requiring high sensitivity and multiplex capabilities.

  • Product type: Multiplex immunoassay kit
  • Species reactivity: Human
  • Number of analytes: 10
  • Analytes detected: IFN-γ, IL-1β, IL-2, IL-4, IL-5, IL-8, IL-10, IL-12p70, IL-13, TNF-α
  • Pathway focus: Inflammation, Th1/Th2 pathways
  • Detection technology: Electrochemiluminescence (ECL)
  • Sensitivity: Sub- to low pg/mL range
  • Sample types: Cell culture supernatant, serum, plasma
  • Research use only: Yes
  • Multiplex compatibility: Yes
  • Kit components: Assays fulfilled as a kit

Catalog No. NC3132558 Supplier Meso Scale Discovery Supplier No. K15010K1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The MESO SCALE DISCOVERY U-PLEX TH1/TH2 Combo (human) is a pre-configured multiplex assay kit designed to measure human cytokines involved in inflammation and the Th1/Th2 pathways. This kit features advanced U-PLEX technology that enables the efficient and sensitive detection of multiple analytes in a single sample, making it an ideal choice for research requiring high sensitivity and multiplex capabilities.

  • Product type: Multiplex immunoassay kit
  • Species reactivity: Human
  • Number of analytes: 10
  • Analytes detected: IFN-γ, IL-1β, IL-2, IL-4, IL-5, IL-8, IL-10, IL-12p70, IL-13, TNF-α
  • Pathway focus: Inflammation, Th1/Th2 pathways
  • Detection technology: Electrochemiluminescence (ECL)
  • Sensitivity: Sub- to low pg/mL range
  • Sample types: Cell culture supernatant, serum, plasma
  • Research use only: Yes
  • Multiplex compatibility: Yes
  • Kit components: Assays fulfilled as a kit

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.