Learn More
Meso Scale Discovery U-PLEX TH1/TH2 Combo NHP
SDP
Catalog No. 5000078812
Encompass
The U-PLEX TH1/TH2 Combo NHP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously measures the TH1 cytokines IFN-gamma IL-2 and TNF-alpha as well as the TH2 cytokines IL-4 IL-5 IL-10 and IL-13 in non-human primate serum plasma and culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes all necessary reagents and calibrators for quantitative analysis of the 7 cytokines The assay has high sensitivity requires a low sample volume and provides results in just a few hours
Catalog No. 50-000-78812 Supplier Meso Scale Discovery Supplier No. K15080K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH1/TH2 Combo NHP Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously measures the TH1 cytokines IFN-gamma IL-2 and TNF-alpha as well as the TH2 cytokines IL-4 IL-5 IL-10 and IL-13 in non-human primate serum plasma and culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes all necessary reagents and calibrators for quantitative analysis of the 7 cytokines The assay has high sensitivity requires a low sample volume and provides results in just a few hours