Learn More
Meso Scale Discovery U-PLEX TH1/TH2 Combo NHP
SDP
Catalog No. 5000081760
Encompass
The U-PLEX TH1/TH2 Combo NHP Assay is a multiplex immunoassay panel that simultaneously measures 10 biomarkers related to T helper cell activation in non-human primate samples It uses electrochemiluminescence detection technology on the Meso Scale Discovery platform to quantitatively measure IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IL-17A TNF- and GM-CSF with high sensitivity and dynamic range from a small sample volume This assay provides a comprehensive profile of key cytokines involved in TH1 and TH2 immune responses for immunology and inflammation research in non-human primate models
Catalog No. 50-000-81760 Supplier Meso Scale Discovery Supplier No. K15080K1
The U-PLEX TH1/TH2 Combo NHP Assay is a multiplex immunoassay panel that simultaneously measures 10 biomarkers related to T helper cell activation in non-human primate samples It uses electrochemiluminescence detection technology on the Meso Scale Discovery platform to quantitatively measure IFN- IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IL-17A TNF- and GM-CSF with high sensitivity and dynamic range from a small sample volume This assay provides a comprehensive profile of key cytokines involved in TH1 and TH2 immune responses for immunology and inflammation research in non-human primate models