Learn More
Meso Scale Discovery U-PLEX TH17 Combo 1 NHP
SDP
Catalog No. 5000080549
Encompass
The U-PLEX TH17 Combo 1 NHP is a multiplex assay kit by Meso Scale Diagnostics that allows simultaneous measurement of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 sCD40L and TNF-alpha from cell culture supernatants serum plasma (citrate heparin) and saliva It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It is compatible with MESO s SECTOR Imager and QuickPlex SQ 120 instruments for analysis
This item is not returnable. View return policy
Catalog No. 50-000-80549 Supplier Meso Scale Discovery Supplier No. K15079K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH17 Combo 1 NHP is a multiplex assay kit by Meso Scale Diagnostics that allows simultaneous measurement of IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 IL-33 sCD40L and TNF-alpha from cell culture supernatants serum plasma (citrate heparin) and saliva It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators It is compatible with MESO s SECTOR Imager and QuickPlex SQ 120 instruments for analysis