Learn More
Meso Scale Discovery U-PLEX TH17 COMBO 1HU SECTOR, multiplex immunoassay kit, 2 plates, 96-well, human Th17 pathway biomarkers
SDP
Catalog No. NC3653317
Encompass
The U-PLEX TH17 Combo 1 is a multiplex immunoassay kit designed for the quantitative measurement of key biomarkers associated with the human Th17 pathway, offering high-sensitivity detection and versatility for various research applications in immunology and inflammation. It provides a complete kit on two 96-well plates for optimal assay performance. This product is intended strictly for research use and not for diagnostic procedures.
  • Comprehensively quantifies 9 critical human Th17 pathway biomarkers in a single assay.
  • Achieves detection limits from sub- to low pg/mL for robust biomarker detection.
  • Convenient, pre-tested panel of assays ensures multiplex compatibility.
  • Optimized for use with serum, plasma, and cell culture supernatant for sample flexibility.
  • Delivered on two separate plates to prevent assay interference and ensure high data quality.
Catalog No. NC3653317 Supplier Meso Scale Discovery Supplier No. K15075K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH17 Combo 1 is a multiplex immunoassay kit designed for the quantitative measurement of key biomarkers associated with the human Th17 pathway, offering high-sensitivity detection and versatility for various research applications in immunology and inflammation. It provides a complete kit on two 96-well plates for optimal assay performance. This product is intended strictly for research use and not for diagnostic procedures.
  • Comprehensively quantifies 9 critical human Th17 pathway biomarkers in a single assay.
  • Achieves detection limits from sub- to low pg/mL for robust biomarker detection.
  • Convenient, pre-tested panel of assays ensures multiplex compatibility.
  • Optimized for use with serum, plasma, and cell culture supernatant for sample flexibility.
  • Delivered on two separate plates to prevent assay interference and ensure high data quality.