Learn More
Meso Scale Discovery U-PLEX TH17 COMBO 3, multiplex immunoassay kit, mouse, 5 plate pack, sensitivity sub- to low pg/mL, dynamic range up to 5 logs, storage 2-8 °C, research use only
SDP
Catalog No. NC3727639
Encompass
The MESO SCALE DISCOVERY U-PLEX TH17 Combo 3 (mouse) is a multiplex immunoassay kit that quantitatively measures 10 key analytes related to inflammation and immune system regulation in mouse samples. Utilizing proprietary MULTI-ARRAY technology and ECL detection, this kit offers high sensitivity and a wide dynamic range for efficient biomarker profiling.
  • Multiplex immunoassay kit
  • Mouse species reactivity
  • Measures 10 analytes
  • Electrochemiluminescence (ECL) assay principle
  • MULTI-ARRAY technology for detection
  • Applicable in immunology and inflammation research
  • Compatible with serum, EDTA plasma, and cell culture supernatants
  • Sub- to low pg/mL sensitivity
  • Dynamic range of up to 5 logs
  • Includes a 5 plate pack
  • Store at 2-8 °C
  • For research use only
  • Detection limits for analytes range from 0.10 pg/mL to 45 pg/mL
Catalog No. NC3727639 Supplier Meso Scale Discovery Supplier No. K15354K2
Add to Cart
Add to Cart
Request a Quote
The MESO SCALE DISCOVERY U-PLEX TH17 Combo 3 (mouse) is a multiplex immunoassay kit that quantitatively measures 10 key analytes related to inflammation and immune system regulation in mouse samples. Utilizing proprietary MULTI-ARRAY technology and ECL detection, this kit offers high sensitivity and a wide dynamic range for efficient biomarker profiling.
  • Multiplex immunoassay kit
  • Mouse species reactivity
  • Measures 10 analytes
  • Electrochemiluminescence (ECL) assay principle
  • MULTI-ARRAY technology for detection
  • Applicable in immunology and inflammation research
  • Compatible with serum, EDTA plasma, and cell culture supernatants
  • Sub- to low pg/mL sensitivity
  • Dynamic range of up to 5 logs
  • Includes a 5 plate pack
  • Store at 2-8 °C
  • For research use only
  • Detection limits for analytes range from 0.10 pg/mL to 45 pg/mL