Learn More
Meso Scale Discovery U-PLEX TH17 Combo 3 (mouse), 10 analytes, 0.10 - 45 pg/mL detection limits, 96-well plate immunoassay kit, Electrochemiluminescence (ECL) detection
SDP
Catalog No. NC2245274
Encompass
The U-PLEX TH17 Combo 3 is a multiplex immunoassay kit designed for the quantitative measurement of key analytes associated with inflammation and immune system regulation in mouse samples. It utilizes MSD's proprietary MULTI-ARRAY technology with electrochemiluminescence (ECL) detection, ensuring high sensitivity and a broad dynamic range of detection limits. This convenient assay format is intended for research use only and serves as an alternative to other unavailable products.
  • Designed for mouse samples
  • Measures 10 analytes related to inflammation
  • Utilizes electrochemiluminescence detection
  • High sensitivity detection limits ranging from 0.10 to 45 pg/mL
  • Broad linear dynamic range of up to 5 logs
  • Provided in a single 96-well plate format
  • Processing time of approximately 4 hours for up to 10 assays
  • Research use only
This item is not returnable. View return policy
Catalog No. NC2245274 Supplier Meso Scale Discovery Supplier No. K15354K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX TH17 Combo 3 is a multiplex immunoassay kit designed for the quantitative measurement of key analytes associated with inflammation and immune system regulation in mouse samples. It utilizes MSD's proprietary MULTI-ARRAY technology with electrochemiluminescence (ECL) detection, ensuring high sensitivity and a broad dynamic range of detection limits. This convenient assay format is intended for research use only and serves as an alternative to other unavailable products.
  • Designed for mouse samples
  • Measures 10 analytes related to inflammation
  • Utilizes electrochemiluminescence detection
  • High sensitivity detection limits ranging from 0.10 to 45 pg/mL
  • Broad linear dynamic range of up to 5 logs
  • Provided in a single 96-well plate format
  • Processing time of approximately 4 hours for up to 10 assays
  • Research use only