Learn More
Meso Scale Discovery U-PLEX Viral Combo 1 human
SDP
Catalog No. 5000079396
Encompass
The U-PLEX Viral Combo 1 Human Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously measures antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum plasma and whole blood samples This assay kit includes a 96-well plate coated with antigens for the 4 viruses detection antibodies assay diluent wash buffer read buffer and calibrators to quantitatively determine antibody levels The electrochemiluminescent detection allows high sensitivity and a broad dynamic range This kit allows efficient high-throughput testing for antibodies to common respiratory viruses
Catalog No. 50-000-79396 Supplier Meso Scale Discovery Supplier No. K15343K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Viral Combo 1 Human Kit from Meso Scale Diagnostics is a multiplex immunoassay that simultaneously measures antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum plasma and whole blood samples This assay kit includes a 96-well plate coated with antigens for the 4 viruses detection antibodies assay diluent wash buffer read buffer and calibrators to quantitatively determine antibody levels The electrochemiluminescent detection allows high sensitivity and a broad dynamic range This kit allows efficient high-throughput testing for antibodies to common respiratory viruses