Learn More
Meso Scale Discovery U-PLEX Viral Combo 1 NHP
SDP
Catalog No. 5000079397
Encompass
The U-PLEX Viral Combo 1 NHP is a multiplex immunoassay panel that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and other biological samples This assay uses Meso Scale Discovery electrochemiluminescence technology and provides quantitative results in under 3 hours The panel includes antigens for the SARS-CoV-2 spike and nucleocapsid proteins Influenza A nucleoprotein Influenza B nucleoprotein and RSV fusion protein This assay allows researchers to efficiently profile antibody responses to multiple respiratory viruses in non-human primate studies
Catalog No. 50-000-79397 Supplier Meso Scale Discovery Supplier No. K15344K1
The U-PLEX Viral Combo 1 NHP is a multiplex immunoassay panel that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in non-human primate serum plasma and other biological samples This assay uses Meso Scale Discovery electrochemiluminescence technology and provides quantitative results in under 3 hours The panel includes antigens for the SARS-CoV-2 spike and nucleocapsid proteins Influenza A nucleoprotein Influenza B nucleoprotein and RSV fusion protein This assay allows researchers to efficiently profile antibody responses to multiple respiratory viruses in non-human primate studies