Learn More
Meso Scale Discovery U-PLEX Viral Combo 1 NHP
SDP
Catalog No. 5000081886
Encompass
The U-PLEX Viral Combo 1 NHP is a multiplex immunoassay panel from Meso Scale Diagnostics that allows simultaneous quantitative measurement of interferon gamma interleukin 10 interleukin 17A/F interleukin 2 interleukin 4 interleukin 6 and tumor necrosis factor alpha in non-human primate samples such as serum plasma and culture supernatants This 7-plex panel utilizes Meso Scale s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and low sample volume requirements The panel is designed for immunology and inflammation research applications involving non-human primate models
Catalog No. 50-000-81886 Supplier Meso Scale Discovery Supplier No. K15344K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Viral Combo 1 NHP is a multiplex immunoassay panel from Meso Scale Diagnostics that allows simultaneous quantitative measurement of interferon gamma interleukin 10 interleukin 17A/F interleukin 2 interleukin 4 interleukin 6 and tumor necrosis factor alpha in non-human primate samples such as serum plasma and culture supernatants This 7-plex panel utilizes Meso Scale s electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and low sample volume requirements The panel is designed for immunology and inflammation research applications involving non-human primate models